missing translation for 'onlineSavingsMsg'
Learn More

MSH2 Antibody (CL13724) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30525030 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525030 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525030 Supplier Novus Biologicals Supplier No. NBP343852MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL13724
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P43246, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4436
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.