missing translation for 'onlineSavingsMsg'
Learn More

MSH3 Antibody (CL14059) [DyLight 350], Novus Biologicals™

Product Code. 30525125 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525125 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525125 Supplier Novus Biologicals Supplier No. NBP343855UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH3
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL14059
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli)
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P20585, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4437
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.