missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MSH6 Monoclonal specifically detects MSH6 in Ferret/Human/Monkey/Mouse/Rat samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | MSH6 |
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 035 |
| Conjugate | PerCP |
| Dilution | Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human MSH6. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?