missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marke: Novus Biologicals NB100-56483
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MTA2 Polyclonal specifically detects MTA2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| MTA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
| Rabbit | |
| 75 kDa | |
| 0.1 mg | |
| Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
| 9219 | |
| Human, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| O94776 | |
| MTA2 | |
| A portion of amino acids 650-700 of human MTA2 was used as the immunogen (NP_004730). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with Chimpanzee, Human and Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Mouse (94%), Rat (94%), Dog (88%), Bovine (88%) and Xenopus (76%). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur