missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
213.00 € - 351.00 €
Specifications
| Antigen | MTA2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038155
|
Novus Biologicals
NB100-56483SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749603
|
Novus Biologicals
NB100-56483 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTA2 Polyclonal specifically detects MTA2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MTA2 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
| MTA2 | |
| IgG | |
| Affinity Purified | |
| Reacts with Chimpanzee, Human and Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Mouse (94%), Rat (94%), Dog (88%), Bovine (88%) and Xenopus (76%). |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Primate | |
| O94776 | |
| 9219 | |
| A portion of amino acids 650-700 of human MTA2 was used as the immunogen (NP_004730). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 75 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title