missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCO2 Mouse, anti-Human, Clone: 10G14F4, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | MTCO2 |
|---|---|
| Clone | 10G14F4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074235
|
Proteintech
68527-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18019875
|
Proteintech
68527-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MTCO2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human MTCO2. This antibody recognizes human antigen. The MTCO2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MTCO2 | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2 | |
| Primary |
| 10G14F4 | |
| Western Blot, ELISA | |
| Mouse | |
| 4513 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title