missing translation for 'onlineSavingsMsg'
Learn More

MTGR1 Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30510978 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30510978 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510978 Supplier Novus Biologicals Supplier No. NBP338369CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTGR1 Polyclonal antibody specifically detects MTGR1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTGR1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias core-binding factor, runt domain, alpha subunit 2; translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 325-604 of human MTGR1 (NP_005084.1).,, Sequence:, GLNGGYQDELVDHRLTEREWADEWKHLDHALNCIMEMVEKTRRSMAVLRRCQESDREELNYWKRRYNENTELRKTGTELVSRQHSPGSADSLSNDSQREFNSRPGTGYVPVEFWKKTEEAVNKVKIQAMSEVQKAVAEAEQKAFEVIATERARMEQTIADVKRQAAEDAFLVINEQEESTENCWNCGRKASETCSGCNIARYCGSFCQHKDWERHHRLCGQNLHGQSPHGQGRPLLPVGRGSSARSADCSVPSPALDKTSATTSRSSTPASVTAIDTNGL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9139
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.