missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68713
This item is not returnable.
View return policy
Description
MTR Polyclonal antibody specifically detects MTR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MTR | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| 5-methyltetrahydrofolate-homocysteine methyltransferase, 5-methyltetrahydrofolate-homocysteine methyltransferase 1, cblG, cobalamin-dependent methionine synthase, EC 2.1.1.13, FLJ33168, FLJ43216,5-methyltetrahydrofolate--homocysteine methyltransferase, FLJ45386, methionine synthase, MS, Vitamin-B12 dependent methionine synthase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DIGKNIVGVVLGCNNFRVIDLGVMTPCDKILKAALDHKADIIGLSGLITPSLDEMIFVAKEMERLAIRIPLLI | |
| 100 μg | |
| Lipid and Metabolism | |
| 4548 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction