missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 593.00 €
Specifications
| Antigen | MTR |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687156
|
Novus Biologicals
NBP2-68713-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642079
|
Novus Biologicals
NBP2-68713 |
100 μg |
593.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTR Polyclonal antibody specifically detects MTR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MTR | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4548 | |
| IgG | |
| Protein A purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 5-methyltetrahydrofolate-homocysteine methyltransferase, 5-methyltetrahydrofolate-homocysteine methyltransferase 1, cblG, cobalamin-dependent methionine synthase, EC 2.1.1.13, FLJ33168, FLJ43216,5-methyltetrahydrofolate--homocysteine methyltransferase, FLJ45386, methionine synthase, MS, Vitamin-B12 dependent methionine synthase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DIGKNIVGVVLGCNNFRVIDLGVMTPCDKILKAALDHKADIIGLSGLITPSLDEMIFVAKEMERLAIRIPLLI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel