missing translation for 'onlineSavingsMsg'
Learn More

GIT1 Antibody (924640) [Allophycocyanin], R&D Systems™

Product Code. 18613995 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613995 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613995 Supplier Novus Biologicals Supplier No. FAB8508A
  • ONLINE EXCLUSIVE PRICE
    709.02 €
    609.00 € / 0.1mL
    Save 100.02 €
    14% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for CyTOF, ELISA, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIT1
Applications CyTOF, ELISA, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 924640
Conjugate APC
Dilution Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28964
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.