missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVK Antibody (SR1713), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22554-100ul
This item is not returnable.
View return policy
Description
MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| MVK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1713 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
| A synthesized peptide derived from human MVK (Uniprot #: Q03426) | |
| 100 μg | |
| metabolism | |
| 4598 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction