missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVK Antibody (SR1713), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | MVK |
|---|---|
| Clone | SR1713 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660834
|
Novus Biologicals
NBP3-22554-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685274
|
Novus Biologicals
NBP3-22554-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| MVK | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
| A synthesized peptide derived from human MVK (Uniprot #: Q03426) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1713 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4598 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title