missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MYB [p Ser11] Antibody (SR2809), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27715-50ul
This item is not returnable.
View return policy
Description
MYB Monoclonal antibody specifically detects MYB in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| MYB | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4602 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR2809 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| Cmyb;c-myb;c-myb_CDS;efg;Transcriptional activator Myb | |
| A synthesized peptide derived from human MYB (Uniprot #: P10242 ) | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction