missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MYB [p Ser11] Antibody (SR2809), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00 € - 382.00 €
Specifications
| Antigen | MYB |
|---|---|
| Clone | SR2809 |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041739
|
Novus Biologicals™
NBP3-27715-50ul |
50 μL |
316.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041598
|
Novus Biologicals™
NBP3-27715-100ul |
100 μL |
382.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYB Monoclonal antibody specifically detects MYB in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| MYB | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 4602 | |
| IgG | |
| Affinity purified |
| SR2809 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Cmyb;c-myb;c-myb_CDS;efg;Transcriptional activator Myb | |
| A synthesized peptide derived from human MYB (Uniprot #: P10242 ) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title