missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (2C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64803-0.025MG
This item is not returnable.
View return policy
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Myeloperoxidase/MPO | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:1000-1:5000 | |
| P05164 | |
| MPO | |
| Human Myeloperoxidase | |
| 0.025 mg | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 4353 | |
| Human | |
| Purified |
| Immunohistochemistry (Frozen), Flow Cytometry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| 2C7 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64803 recognizes human myeloperoxidase (MPO). MPO is an important component of azurophilic granules in neutrophils, being involved in microbicidal processes. The protein is a multimer of 2 heavy chains (55kD) and two light chains (15kD), the heavy chains being linked by a disulphide bond. NB100-64803 recognizes native MPO in Western blots, and the heavy chain following boiling of the sample. The antibody also recognizes recombinant MPO in Western blots and weakly in ELISA. The antibody may be of value in the study of myeloid cells and myeloid leukaemias by flow cytometry following cell permeablisation. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction