missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (2C7), Novus Biologicals™
Mouse Monoclonal Antibody
222.00 € - 524.00 €
Specifications
| Antigen | Myeloperoxidase/MPO |
|---|---|
| Clone | 2C7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:1000-1:5000 |
| Applications | Immunohistochemistry (Frozen), Flow Cytometry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18094444
|
Novus Biologicals
NB100-64803-0.025MG |
0.025 mg |
222.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710414
|
Novus Biologicals
NB100-64803 |
0.1 mg |
524.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Myeloperoxidase/MPO | |
| 1 mg/mL | |
| Immunohistochemistry (Frozen), Flow Cytometry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| P05164 | |
| 4353 | |
| Human Myeloperoxidase | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2C7 | |
| Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:1000-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| MPO | |
| IgG1 | |
| Protein G purified | |
| NB100-64803 recognizes human myeloperoxidase (MPO). MPO is an important component of azurophilic granules in neutrophils, being involved in microbicidal processes. The protein is a multimer of 2 heavy chains (55kD) and two light chains (15kD), the heavy chains being linked by a disulphide bond. NB100-64803 recognizes native MPO in Western blots, and the heavy chain following boiling of the sample. The antibody also recognizes recombinant MPO in Western blots and weakly in ELISA. The antibody may be of value in the study of myeloid cells and myeloid leukaemias by flow cytometry following cell permeablisation. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title