missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82061-0.025ML
This item is not returnable.
View return policy
Description
Myosin Light Chain 2 Polyclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Myosin Light Chain 2 | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
| Rabbit | |
| 18 kDa | |
| 0.025 mL | |
| Primary | |
| Myosin Light Chain 2 (specific to a.a. 18) detects endogenous levels of total Myosin Light Chain 2 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
| P10916, P08733 | |
| MYL2 | |
| The antiserum was produced against synthesized non-phosphopeptide derived from mouse Myosin Light Chain 2 around aa. 18 (A-T-S-N-V). | |
| Affinity Purified | |
| RUO | |
| 4633 | |
| Rat, Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction