missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 486.00 €
Specifications
| Concentration | 1.0 mg/mL |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227105
|
Novus Biologicals
NB100-82061-0.025ML |
0.025 mL |
189.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705733
|
Novus Biologicals
NB100-82061 |
486.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Myosin Light Chain 2 Polyclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| 1.0 mg/mL | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| P10916, P08733 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
| Polyclonal | |
| Rabbit | |
| Rat, Human, Mouse | |
| IgG | |
| Affinity Purified | |
| 18 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title