missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Myogenin Antibody (F12B), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30816-25ug
This item is not returnable.
View return policy
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Gel Super Shift Assays,Immunocytochemistry/ Immunofluorescence
Specifications
| Myogenin | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Gel Supershift, Immunocytochemistry/Immunofluorescence | |
| F12B | |
| Western Blot : 1 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg, Immunohistochemistry-Paraffin : 1-2 μg/mL, Gel Super Shift Assays | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with recombinant protein containing rat myogenin amino acids 30-224 and mouse myeloma NS1 cells. | |
| 25 μg | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction