missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Myogenin Antibody (F12B), Novus Biologicals™
Mouse Monoclonal Antibody
213.00 € - 713.00 €
Specifications
| Antigen | Myogenin |
|---|---|
| Clone | F12B |
| Dilution | Western Blot : 1 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg, Immunohistochemistry-Paraffin : 1-2 μg/mL, Gel Super Shift Assays |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Gel Supershift, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066836
|
Novus Biologicals™
NBP3-30816-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067140
|
Novus Biologicals™
NBP3-30816-100ug |
100 μg |
713.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Gel Super Shift Assays,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Myogenin | |
| Western Blot : 1 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg, Immunohistochemistry-Paraffin : 1-2 μg/mL, Gel Super Shift Assays | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with recombinant protein containing rat myogenin amino acids 30-224 and mouse myeloma NS1 cells. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| F12B | |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Gel Supershift, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| Phosphate buffered saline | |
| 4656 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title