missing translation for 'onlineSavingsMsg'
Learn More

Myozenin 2 Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30508244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30508244 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30508244 Supplier Novus Biologicals Supplier No. NBP338274AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Myozenin 2 Polyclonal antibody specifically detects Myozenin 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myozenin 2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias C4orf5, calcineurin-binding protein calsarcin-1, Calsarcin-1, chromosome 4 open reading frame 5, CS-1, FATZ related protein 2, FATZ-related protein 2, muscle-specific protein, myozenin 2, myozenin-2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-264 of human Myozenin 2 (NP_057683.1).,, Sequence:, MLSHNTMMKQRKQQATAIMKEVHGNDVDGMDLGKKVSIPRDIMLEELSHLSNRGARLFKMRQRRSDKYTFENFQYQSRAQINHSIAMQNGKVDGSNLEGGSQQAPLTPPNTPDPRSPPNPDNIAPGYSGPLKEIPPEKFNTTAVPKYYQSPWEQAISNDPELLEALYPKLFKPEGKAELPDYRSFNRVATPFGGFEKASRMVKFKVPDFELLLLTDPRFMSFVNPLSGRRSFNRTPKGWISENIPIVITTEPTDDTTVPESEDL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51778
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.