missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYRF Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05627-100ul
This item is not returnable.
View return policy
Description
MYRF Polyclonal antibody specifically detects MYRF in Human, Mouse samples. It is validated for Western Blot
Specifications
| MYRF | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 11orf9, C11orf9, CUGS, MMERV, MRF, myelin regulatory factor, Ndt80, pqn-47 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MYRF (NP_001120864.1). MEVVDETEALQRFFEGHDINGALEPSNIDTSILEEYISKEDASDLCFPDISAPASSASYSHGQPAMPGSSGVHHLSPPGGGPSPGRHGPLPPPGYGTPLN | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 745 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction