missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYRF Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00 € - 498.00 €
Specifications
| Antigen | MYRF |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682598
|
Novus Biologicals
NBP3-05627-100ul |
100 μg |
498.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631858
|
Novus Biologicals
NBP3-05627-20ul |
20 μg |
214.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYRF Polyclonal antibody specifically detects MYRF in Human, Mouse samples. It is validated for Western BlotSpecifications
| MYRF | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| 11orf9, C11orf9, CUGS, MMERV, MRF, myelin regulatory factor, Ndt80, pqn-47 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MYRF (NP_001120864.1). MEVVDETEALQRFFEGHDINGALEPSNIDTSILEEYISKEDASDLCFPDISAPASSASYSHGQPAMPGSSGVHHLSPPGGGPSPGRHGPLPPPGYGTPLN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 745 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title