missing translation for 'onlineSavingsMsg'
Learn More

MZF1 Antibody (PCRP-MZF1-1E8), Novus Biologicals™

Product Code. 18667944 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667944 20 μg 20µg
18657534 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18667944 Supplier Novus Biologicals Supplier No. NBP32384920ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MZF1 Monoclonal antibody specifically detects MZF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MZF1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-MZF1-1E8
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias myeloid zinc finger 1, MZF1B, MZF-1zinc finger protein 42 (myeloid-specific retinoic acid-responsive), Zfp98, Zinc finger protein 42, ZNF42MZF, ZSCAN6Zinc finger and SCAN domain-containing protein 6
Host Species Mouse
Immunogen Recombinant fragment (around aa37-128) of human MZF1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7593
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.