missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)-1-naphthylacetamide, TRC

Product Code. 30136856
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136856 1 g 1g
30009579 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136856 Supplier Toronto Research Chemicals Supplier No. TRCA6091401G
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36321-43-4
Molecular Formula C14 H16 N2 O
Molecular Weight (g/mol) 228.29
Synonym N-(2-Aminoethyl)-1-Naphthaleneacetamide, N-(2-Aminoethyl)-1-naphthylacetamide, Naphazoline Imp. A (EP), N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide, Naphthylacetylethylenediamine
IUPAC Name N-(2-aminoethyl)-2-naphthalen-1-ylacetamide
SMILES NCCNC(=O)Cc1cccc2ccccc12

Specifications

Analyte or Component Names Naphthylacetylethylenediamine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H16N2O/c15-8-9-16-14(17)10-12-6-3-5-11-4-1-2-7-13(11)12/h1-7H,8-10,15H2,(H,16,17)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 228.1263
Percent Purity >95%
Chemical Name or Material Naphthylacetylethylenediamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.