missing translation for 'onlineSavingsMsg'
Learn More

N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride, Mikromol™

Product Code. 30051196
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051196 100 mg 100mg
30050757 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051196 Supplier Mikromol Supplier No. MM0487.05
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride
CAS 482577-59-3
Analyte or Component Names N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H22 N2 O2 . Cl H
Certifications/Compliance ISO 17025
Synonym L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-, methyl ester, hydrochloride (1:1), : L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-, methyl ester, monohydrochloride (9CI), N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine methyl ester hydrochloride (1:1)
InChI Formula InChI=1S/C20H22N2O2.ClH/c1-14(2)19(20(23)24-3)22-13-15-8-10-16(11-9-15)18-7-5-4-6-17(18)12-21;/h4-11,14,19,22H,13H2,1-3H3;1H/t19-;/m0./s1
SMILES Cl.COC(=O)[C@@H](NCc1ccc(cc1)c2ccccc2C#N)C(C)C
IUPAC Name methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 358.86
Quantity 100 mg
Formula Weight 358.1448 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.