missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine, TRC

Product Code. 30003887
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003887 10 mg 10mg
30003749 5 mg 5mg
30011518 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003887 Supplier Toronto Research Chemicals Supplier No. TRCE92651010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1418095-19-8
Molecular Formula C14 H19 N O6 S
Molecular Weight (g/mol) 329.37
Synonym (S)-2-(N-(2-Ethyl-6-methylphenyl)-2-sulfoacetamido)propanoic Acid, N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine
IUPAC Name (2S)-2-(2-ethyl-6-methyl-N-(2-sulfoacetyl)anilino)propanoic acid
SMILES CCc1cccc(C)c1N([C@@H](C)C(=O)O)C(=O)CS(=O)(=O)O

Specifications

Analyte or Component Names N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H19NO6S/c1-4-11-7-5-6-9(2)13(11)15(10(3)14(17)18)12(16)8-22(19,20)21/h5-7,10H,4,8H2,1-3H3,(H,17,18)(H,19,20,21)/t10-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 329.0933
Chemical Name or Material N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.