missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenyl]thiourea, 97%, Thermo Scientific™

Product Code. 11378435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378435 1 g 1g
11388435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378435 Supplier Thermo Scientific Alfa Aesar Supplier No. L10444.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-70-5
Molecular Formula C8H7F3N2S
Molecular Weight (g/mol) 220.21
MDL Number MFCD00041171
InChI Key WKRUQAYFMKZMPJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylthiourea, 1-3-trifluoromethyl phenyl thiourea, 3-trifluoromethyl phenyl thiourea, n-3-trifluoromethyl phenyl thiourea, thiourea, 3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl-2-thiourea, amino 3-trifluoromethyl phenyl amino methane-1-thione, acmc-20ah62
PubChem CID 735844
IUPAC Name [3-(trifluoromethyl)phenyl]thiourea
SMILES NC(=S)NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 103°C to 106°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2695477
Formula Weight 220.22
Percent Purity 97%
Chemical Name or Material N-[3-(Trifluoromethyl)phenyl]thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.