missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide, Mikromol

Product Code. 30462411
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30462411 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462411 Supplier Mikromol Supplier No. MM0064.02
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
CAS 41526-21-0
Analyte or Component Names N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
Shipping Condition Room Temperature
Molecular Formula C14 H10 Br Cl N2 O2
Certifications/Compliance ISO 17025
Synonym N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide, Bromazepam Imp. B (EP), Bromazepam Impurity B
InChI Formula InChI=1S/C14H10BrClN2O2/c15-9-4-5-11(18-13(19)8-16)10(7-9)14(20)12-3-1-2-6-17-12/h1-7H,8H2,(H,18,19)
Solution Type Neat
SMILES ClCC(=O)Nc1ccc(Br)cc1C(=O)c2ccccn2
IUPAC Name N-[4-bromo-2-(pyridine-2-carbonyl)phenyl]-2-chloroacetamide
Molecular Weight (g/mol) 353.6
Quantity 50 mg
Formula Weight 351.9614 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.