missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide, Mikromol™

Product Code. 30051737
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051737 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051737 Supplier Mikromol Supplier No. MM0894.180025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
CAS 90357-53-2
Analyte or Component Names N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H9 F3 N2 O
Certifications/Compliance ISO 9001
Synonym 2-Propenamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methyl-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyl-2-propenamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylacrylamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
InChI Formula InChI=1S/C12H9F3N2O/c1-7(2)11(18)17-9-4-3-8(6-16)10(5-9)12(13,14)15/h3-5H,1H2,2H3,(H,17,18)
SMILES CC(=C)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
Molecular Weight (g/mol) 254.2079
Quantity 25 mg
Formula Weight 254.0667 g/mol
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.