missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-cysteine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30327988
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327988 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327988 Supplier Dr. Ehrenstorfer Supplier No. DREA10023150AL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-cysteine
CAS 616-91-1
Analyte or Component Names N-Acetyl-L-cysteine
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym N-Acetyl-L-cysteine, (R)-N-Acetylcysteine, ACC, Acetilcysteina, Acetylcysteine, Airbron, Broncholysin, Broncholysin (mucolytic), Brunac, Exomuc, Fabrol, Fluatox, Fluibiotic, Fluimicil, Fluimicil Infantil, Fluimucetin, Fluimucil, Flumil, Fluprowit, Hypotears, L-Acetylcysteine, L-N-Acetylcysteine, Mercapturic acid, (R)-Mercapturic acid, Muco Sanigen, Mucocedyl, Mucofilin, Mucolator, Mucolyticum, Mucolyticum-Lappe, Mucolytikum Lappe, Mucomyst, Mucosolvin, Mucret, N-Acetyl-(R)-cysteine, N-Acetylcysteine, NSC 111180, Neo-Fluimucil, Parvolex, Respaire, Syntemucol, Tixair, Nalpha-Acetylcysteine
InChI Formula InChI=1S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10H,2H2,1H3,(H,6,7)(H,8,9)/t4-/m0/s1
SMILES CC(=O)N[C@@H](CS)C(=O)O
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 163.19
Quantity 1 mL
Formula Weight 163.0303 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.