missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine, TRC

Product Code. 30179083
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179083 50 g 50g
30179358 10 g 10g
30179495 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179083 Supplier Toronto Research Chemicals Supplier No. TRCA18973550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-55-3
Molecular Formula C11 H13 N O4
Molecular Weight (g/mol) 223.23
Synonym N-Acetyltyrosine, (2S)-2-(Acetylamino)-3-(4-hydroxyphenyl)propanoic acid, Tyrosine, N-acetyl-, L- (6CI,8CI), (+)-(2S)-2-(Acetylamino)-3-(4-hydroxyphenyl)propanoic acid, (2S)-2-Acetylamino-3-(4-hydroxyphenyl)propanoic acid, L-N-Acetyltyrosine, N-Acetyl-L-tyrosine, N-Acetyltyrosine, NSC 10853, Tyr-Excel
IUPAC Name (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O

Specifications

Analyte or Component Names N-Acetyl-L-tyrosine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 g
Formula Weight 223.0845
Chemical Name or Material N-Acetyl-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.