missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylglucosaminyltransferase III/MGAT3 Antibody - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
N-Acetylglucosaminyltransferase III/MGAT3 Polyclonal antibody specifically detects N-Acetylglucosaminyltransferase III/MGAT3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | N-Acetylglucosaminyltransferase III/MGAT3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | beta-1,4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase, EC 2.4.1.144, FLJ43371, GGNT3, GlcNAc-T III, GNT3, GNT-IIIMGC142278, mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, MGC141943, N-acetylglucosaminyltransferase III, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 364-533 of human MGAT3 (NP_002400.3). GCTVDMLQAVYGLDGIRLRRRQYYTMPNFRQYENRTGHILVQWSLGSPLHFAGWHCSWCFTPEGIYFKLVSAQNGDFPRWGDYEDKRDLNYIRGLIRTGGWFDGTQQEYPPADPSEHMYAPKYLLKNYDRFHYLLDNPYQEPRSTAAGGWRHRGPEGRPPARGKLDEAEV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?