missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-1-phenylmethanamine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Benzyl-1-phenylmethanamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-benzyl-1-phenylmethanamine |
| CAS | 20455-68-9 |
| Analyte or Component Names | N-benzyl-1-phenylmethanamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H15 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Noradrenaline Tartrate Imp. F (EP), Noradrenaline Imp. F (EP), N-(phenylmethyl)benzenemethanamine hydrochloride, Dibenzylamine hydrochloride, Dibenzylammonium chloride, Noradrenaline Imp. F (EP) as Hydrochloride, Noradrenaline Tartrate Impurity F as Hydrochloride, Noradrenaline Impurity F as Hydrochloride |
| InChI Formula | InChI=1S/C14H15N.ClH/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14;/h1-10,15H,11-12H2;1H |
| SMILES | Cl.C(NCc1ccccc1)c2ccccc2 |
| IUPAC Name | N-benzyl-1-phenylmethanamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction