missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Benzyloxycarbonyl)-L-phenylalanine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-(Benzyloxycarbonyl)-L-phenylalanine
Specifications
Specifications
| Chemical Name or Material | N-(Benzyloxycarbonyl)-L-phenylalanine |
| CAS | 1161-13-3 |
| Analyte or Component Names | N-(Benzyloxycarbonyl)-L-phenylalanine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Phenylalanine, N-[(phenylmethoxy)carbonyl]-, Alanine, N-carboxy-3-phenyl-, N-benzyl ester, L- (8CI), N-[(Phenylmethoxy)carbonyl]-L-phenylalanine, (2S)-2-((Benzyloxycarbonyl)amino)-3-phenylpropionic acid, (2S)-2-[(Benzyloxycarbonyl)amino]-3-phenylpropanoic acid, (Benzyloxycarbonyl)phenylalanine, (Carbobenzoxy)phenylalanine, (S)-2-(Benzyloxycarbonylamino)-3-phenyl-propanoic acid, (S)-N-(Benzyloxycarbonyl)phenylalanine, Benzyloxycarbonyl-L-phenylalanine, Carbobenzoxy-L-phenylalanine, L-(Carbobenzoxy)phenylalanine, L-(Carbobenzyloxy)phenylalanine, L-Cbz phenylalanine, N-(Benzyloxycarbonyl)-L-phenylalanine, N-(Carbobenzoxy)phenylalanine, N-(Carbobenzyloxy)-L-phenylalanine, N-(Carbobenzyloxy)phenylalanine, N-Benzyloxycarbonyl-(S)-phenylalanine, N-Carbobenzoxy-L-phenylalanine, N-Carboxy-3-phenyl-L-alanine N-benzyl ester, N-Cbz-L-phenylalanine, N-[(Benzyloxy)carbonyl]phenylalanine, N-(Benzyloxycarbonyl)-L-phenylalanine |
| InChI Formula | InChI=1S/C17H17NO4/c19-16(20)15(11-13-7-3-1-4-8-13)18-17(21)22-12-14-9-5-2-6-10-14/h1-10,15H,11-12H2,(H,18,21)(H,19,20)/t15-/m0/s1 |
| SMILES | OC(=O)[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2 |
| IUPAC Name | (2S)-2-(benzyloxycarbonylamino)-3-phenyl-propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction