missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-bromo-3-fluoroaniline, 96%, Thermo Scientific Chemicals

Product Code. 15475698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475698 1 g 1g
15485698 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475698 Supplier Thermo Scientific Alfa Aesar Supplier No. H59398.03
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 868735-43-7 | Molecular Formula: C11H13BrFNO2 | Molecular Weight: 290.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868735-43-7
Molecular Formula C11H13BrFNO2
Molecular Weight (g/mol) 290.132
MDL Number MFCD11976284
InChI Key FQLYUJLJZFATNI-UHFFFAOYSA-N
Synonym tert-butyl 4-bromo-3-fluorophenyl carbamate, tert-butyl n-4-bromo-3-fluorophenyl carbamate, tert-butyl 4-bromo-3-fluorophenylcarbamate, 4-bromo-3-fluoro-phenyl-carbamic acid tert-butyl ester, n-boc-4-bromo-3-fluoroaniline
PubChem CID 59852943
IUPAC Name tert-butyl N-(4-bromo-3-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC(=C(C=C1)Br)F

Specifications

Melting Point 68°C to 70°C
Quantity 1 g
UN Number UN2811
Beilstein 14460051
Formula Weight 290.13
Percent Purity 96%
Chemical Name or Material N-Boc-4-bromo-3-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.