missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-leucine, Mikromol™

Product Code. 17937674
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17937674 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17937674

Brand: Mikromol MM1219.180025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Formyl-L-leucine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl-L-leucine
CAS 6113-61-7
Analyte or Component Names N-Formyl-L-leucine
Shipping Condition Room Temperature
Molecular Formula C7 H13 N O3
Certifications/Compliance ISO 17025
Synonym L-Leucine, N-formyl-, Leucine, N-formyl-, L- (8CI), N-Formyl-L-leucine, (2S)-2-Formamido-4-methylpentanoic acid, (S)-(+)-2-Formamido-4-methylpentanoic acid, N-Formyl-S-leucine, N-Formylleucine, NSC 334321
InChI Formula InChI=1S/C7H13NO3/c1-5(2)3-6(7(10)11)8-4-9/h4-6H,3H2,1-2H3,(H,8,9)(H,10,11)/t6-/m0/s1
SMILES CC(C)C[C@H](NC=O)C(=O)O
IUPAC Name (2S)-2-formamido-4-methylpentanoic acid
Molecular Weight (g/mol) 159.18
Quantity 25 mg
Formula Weight 159.0895 g/mol
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.