missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide, 98%

Product Code. 15410588
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15410588

missing translation for 'mfr': Thermo Scientific Alfa Aesar H54253.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6919-61-5 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6919-61-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00075320
InChI Key UKERDACREYXSIV-UHFFFAOYSA-N
Synonym n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
PubChem CID 569575
ChEBI CHEBI:59742
IUPAC Name N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=CC=C1)OC

Specifications

Density 1.085
Boiling Point 70°C
Flash Point 113°C (235°F)
Refractive Index 1.533
Quantity 25 g
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material N-Methoxy-N-methylbenzamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.