missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmethanamine Hydrochloride, Mikromol™

Product Code. 17949289
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949289 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17949289 Supplier Mikromol Supplier No. MM0056.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Methylmethanamine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methylmethanamine Hydrochloride
CAS 506-59-2
Analyte or Component Names N-Methylmethanamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C2 H7 N . Cl H
Certifications/Compliance ISO 9001
Synonym Dacarbazine Imp. D (EP), Metformin Hydrochloride Imp. F (EP), Metformin Imp. F (EP), N-Methylmethanamine Hydrochloride, Dacarbazine Imp. D (EP) as Hydrochloride, Metformin Hydrochloride Imp. F (EP) as Hydrochloride, Imatinib Mesilate Imp. I (Pharmeuropa), Dimethylamine Hydrochloride, Dacarbazine Impurity D as Hydrochloride, Metformin Hydrochloride Impurity F as Hydrochloride, Metformin Impurity F as Hydrochloride
InChI Formula InChI=1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H
SMILES Cl.CNC
IUPAC Name N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 81.54
Quantity 100 mg
Formula Weight 81.0345 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.