missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline, Mikromol™

Product Code. 30463857
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463857 4 x 25 mg -
30051431 25 mg 25mg
30050315 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463857 Supplier Mikromol Supplier No. MM0378.0600254AMP
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline
CAS 51765-72-1
Analyte or Component Names N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline
Shipping Condition Room Temperature
Molecular Formula C14 H14 N2 O7 S2
Certifications/Compliance ISO 17025
Synonym N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline, Nimesulide Imp. F (EP), Nimesulide Impurity F
InChI Formula InChI=1S/C14H14N2O7S2/c1-24(19,20)16(25(2,21)22)13-9-8-11(15(17)18)10-14(13)23-12-6-4-3-5-7-12/h3-10H,1-2H3
Solution Type Neat
SMILES CS(=O)(=O)N(c1ccc(cc1Oc2ccccc2)[N+](=O)[O-])S(=O)(=O)C
IUPAC Name N-methylsulfonyl-N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
Molecular Weight (g/mol) 386.4
Quantity 4 x 25 mg
Formula Weight 386.0242 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.