missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylethane-1,2-diamine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
N,N-Diethylethane-1,2-diamine
Specifications
Specifications
| Chemical Name or Material | N,N-diethylethane-1,2-diamine |
| CAS | 100-36-7 |
| Analyte or Component Names | N,N-diethylethane-1,2-diamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H16 N2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Metoclopramide Imp. E (EP), Tiapride Hydrochloride Imp. C (EP), Tiapride Imp. C (EP), N,N-Diethylethane-1,2-diamine, Metoclopramide Hydrochloride Monohydrate Impurity E, Metoclopramide Impurity E, Tiapride Hydrochloride Impurity C, Metoclopramide Impurity E, Tiapride Impurity C |
| InChI Formula | InChI=1S/C6H16N2/c1-3-8(4-2)6-5-7/h3-7H2,1-2H3 |
| SMILES | CCN(CC)CCN |
| IUPAC Name | N',N'-diethylethane-1,2-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction