missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-3-chloropropylamine Hydrochloride, Mikromol™

Product Code. 17973146
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973146 25 mg 25mg
17993596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17973146 Supplier Mikromol Supplier No. MM0290.020025
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N,N-Dimethyl-3-chloropropylamine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyl-3-chloropropylamine Hydrochloride
CAS 4-5-5407
Analyte or Component Names N,N-Dimethyl-3-chloropropylamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C5 H12 Cl N . Cl H
Certifications/Compliance ISO 9001
Synonym N,N-Dimethyl-3-chloropropylamine Hydrochloride, 1-Chloropropyl(dimethyl)amine Hydrochloride, Benzydamine Hydrochloride Impurity G, Benzydamine Impurity G
InChI Formula InChI=1S/C5H12ClN.ClH/c1-7(2)5-3-4-6;/h3-5H2,1-2H3;1H
SMILES Cl.CN(C)CCCCl
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 158.07
Quantity 25 mg
Formula Weight 157.0425 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.