missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylpyridin-4-amine, Mikromol™

Product Code. 30050942
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050942 25 mg 25mg
18003920 100 mg 100mg
30458998 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050942 Supplier Mikromol Supplier No. MM0619.070025
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N,N-Dimethylpyridin-4-amine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Dimethylaminopyridine
CAS 1122-58-3
Analyte or Component Names 4-Dimethylaminopyridine
Shipping Condition Room Temperature
Molecular Formula C7 H10 N2
Certifications/Compliance ISO 17025
Synonym Valaciclovir Hydrochloride Imp. G (EP), Valaciclovir Imp. G (EP), Valacyclovir USP Related Compound G, Valacyclovir USP RC G, N,N-Dimethylpyridin-4-amine, Valaciclovir Hydrochloride, Anhydrous Imp. G (EP), Valacyclovir Related Compound G, Valaciclovir Hydrochloride Impurity G, Valaciclovir Impurity G
InChI Formula InChI=1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3
SMILES CN(C)c1ccncc1
IUPAC Name N,N-dimethylpyridin-4-amine
Molecular Weight (g/mol) 122.17
Quantity 25 mg
Formula Weight 122.0844 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.