missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Carteolol, TRC

Product Code. 30416939
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416939 100 mg 100mg
30416940 50 mg 50mg
30426781 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416939 Supplier Toronto Research Chemicals Supplier No. TRCN703620100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Carteolol
Analyte or Component Names N-Nitroso Carteolol
Shipping Condition Room Temperature
Molecular Formula C16 H23 N3 O4
Synonym N-tert-Butyl-N-[2-hydroxy-3-[(2-oxo-3,4-dihydro-1H-quinolin-5-yl)oxy]propyl]nitrous Amide, N-(tert-butyl)-N-(2-hydroxy-3-((2-oxo-1,2,3,4-tetrahydroquinolin-5-yl)oxy)propyl)nitrous amide
InChI Formula InChI=1S/C16H23N3O4/c1-16(2,3)19(18-22)9-11(20)10-23-14-6-4-5-13-12(14)7-8-15(21)17-13/h4-6,11,20H,7-10H2,1-3H3,(H,17,21)
Solution Type Neat
SMILES O=NN(C(C)(C)C)CC(O)COC1=CC=CC2=C1CCC(N2)=O
IUPAC Name N-tert-butyl-N-[2-hydroxy-3-[(2-oxo-3,4-dihydro-1H-quinolin-5-yl)oxy]propyl]nitrous amide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 321.3715
Quantity 100 mg
Formula Weight 321.1689
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.