missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso Desmethyl Almotriptan, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Desmethyl Almotriptan |
| Analyte or Component Names | N-Nitroso Desmethyl Almotriptan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H22 N4 O3 S |
| Synonym | N-Methyl-N-(2-(5-((pyrrolidin-1-ylsulfonyl)methyl)-1H-indol-3-yl)ethyl)nitrous Amide |
| InChI Formula | InChI=1S/C16H22N4O3S/c1-19(18-21)9-6-14-11-17-16-5-4-13(10-15(14)16)12-24(22,23)20-7-2-3-8-20/h4-5,10-11,17H,2-3,6-9,12H2,1H3 |
| Solution Type | Neat |
| SMILES | CN(CCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12)N=O |
| IUPAC Name | N-methyl-N-[2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]ethyl]nitrous amide |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?