missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso Mirabegron, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Mirabegron |
| CAS | 3017159-61-1 |
| Analyte or Component Names | N-Nitroso Mirabegron |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H23 N5 O3 S |
| Synonym | (R)-2-(2-aminothiazol-4-yl)-N-(4-(2-((2-hydroxy-2-phenylethyl)(nitroso)amino)ethyl)phenyl)acetamide |
| InChI Formula | InChI=1S/C21H23N5O3S/c22-21-24-18(14-30-21)12-20(28)23-17-8-6-15(7-9-17)10-11-26(25-29)13-19(27)16-4-2-1-3-5-16/h1-9,14,19,27H,10-13H2,(H2,22,24)(H,23,28)/t19-/m0/s1 |
| Solution Type | Neat |
| SMILES | O=C(NC1=CC=C(CCN(C[C@H](O)C2=CC=CC=C2)N=O)C=C1)CC3=CSC(N)=N3 |
| IUPAC Name | (R)-2-(2-aminothiazol-4-yl)-N-(4-(2-((2-hydroxy-2-phenylethyl)(nitroso)amino)ethyl)phenyl)acetamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?