missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-di-n-butylamine 1.0 mg/ml in Methanol, Mikromol

Product Code. 30458967
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458967 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458967 Supplier Mikromol Supplier No. MM0487.15L21000
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitroso-N-di-n-butylamine 1.0 mg/ml in Methanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-di-n-butylamine
CAS 924-16-3
Analyte or Component Names N-Nitroso-di-n-butylamine
Shipping Condition Ice pack (-20°C)
Molecular Formula C8 H18 N2 O
Concentration 1 mg/mL
Certifications/Compliance ISO 9001
Synonym 1-Butanamine, N-butyl-N-nitroso-, Dibutylamine, N-nitroso- (6CI,7CI,8CI), N-Butyl-N-nitroso-1-butanamine, Di-n-butylnitrosamine, Dibutylnitrosamine, N,N-Di-n-butylnitrosamine, N,N-Dibutylnitrosamine, N-Nitroso-N-di-n-butylamine, N-Nitrosodi-n-butylamine, N-Nitrosodibutylamine, NDBA, NSC 6830, Nitrosodibutylamine, N-Nitroso-di-n-butylamine
InChI Formula InChI=1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3
Solution Type Single Solution
SMILES CCCCN(CCCC)N=O
IUPAC Name N,N-dibutylnitrous amide
Solvent or Matrix Methanol
Molecular Weight (g/mol) 158.24
Quantity 1 mL
Formula Weight 158.1419 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.