missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosoadrenalone, TRC

Product Code. 30477510
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477510 5 mg 5mg
30475891 25 mg 25mg
30466628 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477510 Supplier Toronto Research Chemicals Supplier No. TRCN3447305MG
 Restricted Item
Estimated Shipment: 19-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosoadrenalone
Analyte or Component Names N-Nitrosoadrenalone
Shipping Condition Room Temperature
Molecular Formula C9 H10 N2 O4
Synonym N-[2-(3,4-dihydroxyphenyl)-2-oxo-ethyl]-N-methyl-nitrous amide, N-Nitrosoadrenalone
InChI Formula InChI=1S/C9H10N2O4/c1-11(10-15)5-9(14)6-2-3-7(12)8(13)4-6/h2-4,12-13H,5H2,1H3
Solution Type Neat
SMILES CN(CC(=O)c1ccc(O)c(O)c1)N=O
IUPAC Name N-[2-(3,4-dihydroxyphenyl)-2-oxoethyl]-N-methylnitrous amide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 210.187
Quantity 5 mg
Formula Weight 210.064
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.