missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodi-(n-butyl)amine, Mikromol™

Product Code. 18063100
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063100 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18063100 Supplier Mikromol Supplier No. MM0487.150100
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosodi-(n-butyl)amine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-di-n-butylamine
CAS 924-16-3
Analyte or Component Names N-Nitroso-di-n-butylamine
Shipping Condition Room Temperature
Molecular Formula C8 H18 N2 O
Certifications/Compliance ISO 9001
Synonym 1-Butanamine, N-butyl-N-nitroso-, Dibutylamine, N-nitroso- (6CI,7CI,8CI), N-Butyl-N-nitroso-1-butanamine, Di-n-butylnitrosamine, Dibutylnitrosamine, N,N-Di-n-butylnitrosamine, N,N-Dibutylnitrosamine, N-Nitroso-N-di-n-butylamine, N-Nitrosodi-n-butylamine, N-Nitrosodibutylamine, NDBA, NSC 6830, Nitrosodibutylamine, N-Nitroso-di-n-butylamine
InChI Formula InChI=1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3
SMILES CCCCN(CCCC)N=O
IUPAC Name N,N-dibutylnitrous amide
Molecular Weight (g/mol) 158.24
Quantity 100 mg
Formula Weight 158.1419 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.