missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiisobutylamine, Mikromol

Product Code. 30460406
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30460406 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460406 Supplier Mikromol Supplier No. MM3987.100100
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosodiisobutylamine, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosodiisobutylamine
CAS 997-95-5
Analyte or Component Names N-Nitrosodiisobutylamine
Shipping Condition Room Temperature
Molecular Formula C8 H18 N2 O
Certifications/Compliance ISO 17025
Synonym Dipropylamine, 2,2'-dimethyl-N-nitroso- (6CI,7CI,8CI), 2-Methyl-N-(2-methylpropyl)-N-nitroso-1-propanamine, Diisobutylnitrosamine, Diisobutylnitrosoamine, N,N-Diisobutylnitrosamine, N-Nitroso-di-iso-butylamine, N-Nitrosodiisobutylamine, NSC 134
InChI Formula InChI=1S/C8H18N2O/c1-7(2)5-10(9-11)6-8(3)4/h7-8H,5-6H2,1-4H3
Solution Type Neat
SMILES CC(C)CN(CC(C)C)N=O
IUPAC Name N,N-bis(2-methylpropyl)nitrous amide
Molecular Weight (g/mol) 158.24
Quantity 100 mg
Formula Weight 158.1419 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.